Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METRN, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89131862 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-131-862 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-131-862 Supplier Abnova Corporation Supplier No. PAB5450
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of METRN.

Meteorin regulates glial cell differentiation and promotes the formation of axonal networks during neurogenesis (Nishino et al., 2004 [PubMed 15085178]).[supplied by OMIM

Specifications

Antigen METRN
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of METRN.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene METRN
Gene Alias C16orf23/MGC2601/c380A1.2
Gene Symbols METRN
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human METRN.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79006
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.