Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTL18, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89124385 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-124-385 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-124-385 Supplier Abnova Corporation Supplier No. PAB22676

Rabbit polyclonal antibody raised against recombinant METTL18.

Sequence: NKVIETLPGFQHVKLSVVKTILLKENFPGENIVSKSFSSHSDLITGVYEGGLKIWECTFDLLAYFTKAKVKFAGKKVLDLGCGSGL

Specifications

Antigen METTL18
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description methyltransferase like 18
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene METTL18
Gene Alias RP1-117P20.4, AsTP2, C1orf156, HPM1
Gene Symbols METTL18
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human METTL18.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92342
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.