Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKI67 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114496 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-496 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-496 Supplier Abnova Corporation Supplier No. PAB0889

Rabbit polyclonal antibody raised against a synthetic peptide of MKI67.

This gene encodes a nuclear protein that is associated with and may be necessary for cellular proliferation. Alternatively spliced transcript variants have been described. A related pseudogene exists on chromosome X. [provided by RefSeq

Specifications

Antigen MKI67
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description antigen identified by monoclonal antibody Ki-67
Formulation In PBS (0.2% gelatin, 0.09% sodium azide)
Gene MKI67
Gene Alias KIA, Ki-67
Gene Symbols MKI67
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human MKI67.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4288
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.