Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLF1IP Rabbit anti-Bovine, Chicken, Chimpanzee, Dog, Horse, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114737 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-737 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-737 Supplier Abnova Corporation Supplier No. PAB10267
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant MLF1IP.

The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA (MIM 117139) replaces histone H3 (see MIM 601128). MLF1IP, or CENPU, is an additional factor required for centromere assembly (Foltz et al., 2006 [PubMed 16622419]).[supplied by OMIM

Specifications

Antigen MLF1IP
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant MLF1IP.
Dilution ELISA (1:650000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene MLF1IP
Gene Alias CENP-50/CENP-U/CENP-U(50)/CENP50/CENPU/FLJ23468/KLIP1/PBIP1
Gene Symbols MLF1IP
Host Species Rabbit
Immunogen Recombinant protein corresponding to C-terminus 418 amino acids of human MLF1IP.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79682
Test Specificity This affinity purified antibody is directed against human MLF1IP protein.
Target Species Bovine, Canine, Chicken, Chimpanzee, Equine, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.