Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLIP, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MLIP.
Supplier: Abnova Corporation PAB25793
Specifications
MLIP | |
Polyclonal | |
Unconjugated | |
Western Blot (1 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
MLIP | |
MLIP | |
A synthetic peptide corresponding to 16 amino acids at C-terminus of human MLIP. | |
100 μg | |
Primary | |
At least three isoforms of MLIP are known to exist. | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of MLIP. | |
In PBS (0.02% sodium azide) | |
RP11-642N5.1/C6orf142 | |
Rabbit | |
Peptide affinity purification | |
RUO | |
90523 | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction