Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLL (phospho S516), Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89117954 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-954 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-954 Supplier Abnova Corporation Supplier No. PAB15909
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of MLL.

The MLL gene encodes a DNA-binding protein that methylates histone H3 (see MIM 601128) lys4 (H3K4) and positively regulates expression of target genes, including multiple HOX genes (see MIM 142980). MLL is a frequent target for recurrent translocations in acute leukemias that may be characterized as acute myeloid leukemia (AML; MIM 601626), acute lymphoblastic leukemia (ALL), or mixed lineage (biphenotypic) leukemia (MLL). Leukemias with translocations involving MLL possess unique clinical and biologic characteristics and are often associated with poor prognosis. MLL rearrangements are found in more than 70% of infant leukemias, whether the immunophenotype is more consistent with ALL or AML6, but are less frequent in leukemias from older children. MLL translocations are also found in approximately 10% of AMLs in adults, as well as in therapy-related leukemias, most often characterized as AML, that develop in patients previously treated with topoisomerase II inhibitors for other malignancies. More than 50 different MLL fusion partners have been identified. Leukemogenic MLL translocations encode MLL fusion proteins that have lost H3K4 methyltransferase activity. A key feature of MLL fusion proteins is their ability to efficiently transform hematopoietic cells into leukemia stem cells (Krivtsov and Armstrong, 2007 [PubMed 17957188]).[supplied by OMIM

Specifications

Antigen MLL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of MLL.
Dilution ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene MLL
Gene Alias ALL-1/CXXC7/FLJ11783/HRX/HTRX1/KMT2A/MLL/GAS7/MLL1A/TET1-MLL/TRX1
Gene Symbols MLL
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S516 of human MLL.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4297
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.