Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP23A, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115310 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-310 Supplier Abnova Corporation Supplier No. PAB11706

Rabbit polyclonal antibody raised against synthetic peptide of MMP23A.

This locus represents the partially duplicated copy of the MMP23B gene, and therefore considered a pseudogene. This pseudogene is part of a duplicated region of chromosome 1p36.3. It belongs to the more centromeric copy of the duplicated region. [provided by RefSeq

Specifications

Antigen MMP23A
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration Lot specific
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of MMP23A.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (Formalin/PFA-fixed paraffin-embedded sections) (1:25) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Gene MMP23A
Gene Alias MIFR/MIFR-1/MMP21
Gene Symbols MMP23A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human MMP23A.
Purification Method Immunoaffinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8511
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.