Learn More
MMP8 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MMP8.
Supplier: Abnova Corporation PAB4797
Description
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. However, the enzyme encoded by this gene is stored in secondary granules within neutrophils and is activated by autolytic cleavage. Its function is degradation of type I, II and III collagens. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. [provided by RefSeq
Specifications
MMP8 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of MMP8. | |
In PBS (0.09% sodium azide) | |
CLG1/HNC/PMNL-CL | |
Rabbit | |
Protein G purification | |
RUO | |
4317 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
MMP8 | |
MMP8 | |
A synthetic peptide (conjugated with KLH) corresponding to internal region of human MMP8. | |
400 μL | |
Primary | |
Human, Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only