Learn More
Invitrogen™ Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP
Description
Minimum cross reactivity to human serum proteins.
Specifications
Specifications
| Antigen | Mouse IgG (H+L) Cross-Adsorbed |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 1% BSA and 0.01% gentamicin sulfate; pH 7.2 |
| Host Species | Rabbit |
| Immunogen | Mouse IgG whole molecule. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 100 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.