Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mre11a, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114747 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-747 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-747 Supplier Abnova Corporation Supplier No. PAB10276
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Mre11a.

Specifications

Antigen Mre11a
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Mre11a.
Dilution ELISA (1:10000-1:50000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Mre11a
Gene Alias Mre11/Mre11b
Gene Symbols Mre11a
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 68-706 of mouse Mre11a.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17535
Test Specificity This affinity purified antibody is directed against mouse Mre11 protein.
Target Species Canine, Human, Macaque, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.