Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSH2, Rabbit, Clone: SP46, Abnova™

Catalog No. 89112937 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-937 Supplier Abnova Corporation Supplier No. MAB6556

Rabbit monoclonal antibody raised against synthetic peptide of MSH2.

MSH2 was identified as a locus frequently mutated in hereditary nonpolyposis colon cancer (HNPCC). When cloned, it was discovered to be a human homolog of the E. coli mismatch repair gene mutS, consistent with the characteristic alterations in microsatellite sequences (RER+ phenotype) found in HNPCC. [provided by RefSeq]

Specifications

Antigen MSH2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP46
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of MSH2.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.6 (1% BSA, 0.09% sodium azide).
Gene MSH2
Gene Alias COCA1/FCC1/HNPCC/HNPCC1/LCFS2
Gene Symbols MSH2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region of human MSH2.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4436
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.