Learn More
MSI2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MSI2.
Supplier: Abnova Corporation PAB19416
Description
This gene encodes a protein containing two conserved tandem RNA recognition motifs. Similar proteins in other species function as RNA-binding proteins and play central roles in posttranscriptional gene regulation. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Specifications
MSI2 | |
Polyclonal | |
Unconjugated | |
Western Blot (1 ug/mL) Immunocytochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
MSI2 | |
MSI2 | |
A synthetic peptide corresponding to 15 amino acids near N-terminus of human MSI2. | |
100 μg | |
Primary | |
MSI2 antibody is predicted to not cross-react with MSI1. Multiple isoforms of MSI2 are known to exist. | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunocytochemistry, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of MSI2. | |
In PBS (0.02% sodium azide) | |
FLJ36569/MGC3245/MSI2H | |
Rabbit | |
Peptide affinity purification | |
RUO | |
124540 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only