Learn More
MSR1 Rabbit anti-Bovine, Dog, Horse, Human, Mouse, Primates, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MSR1.
Supplier: Abnova Corporation PAB12379
Description
This gene encodes the class A macrophage scavenger receptors, which include three different types (1, 2, 3) generated by alternative splicing of this gene. These receptors or isoforms are macrophage-specific trimeric integral membrane glycoproteins and have been implicated in many macrophage-associated physiological and pathological processes including atherosclerosis, Alzheimer's disease, and host defense. The isoforms type 1 and type 2 are functional receptors and are able to mediate the endocytosis of modified low density lipoproteins (LDLs). The isoform type 3 does not internalize modified LDL (acetyl-LDL) despite having the domain shown to mediate this function in the types 1 and 2 isoforms. It has an altered intracellular processing and is trapped within the endoplasmic reticulum, making it unable to perform endocytosis. The isoform type 3 can inhibit the function of isoforms type 1 and type 2 when co-expressed, indicating a dominant negative effect and suggesting a mechanism for regulation of scavenger receptor activity in macrophages. [provided by RefSeq]
Specifications
MSR1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of MSR1. | |
In PBS (30% glycerol, 0.09% sodium azide) | |
CD204/SCARA1/SR-A/phSR1/phSR2 | |
Rabbit | |
100 μL | |
Primary | |
Bovine, Canine, Equine, Human, Mouse, Primate, Rat | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Immunocytochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
MSR1 | |
MSR1 | |
A synthetic peptide (linear form) corresponding to amino acids 300-400 of human MSR1. | |
RUO | |
4481 | |
Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only