Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metallothionein, Rabbit, Clone: N11-G, Abnova™

Catalog No. 89113781 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-781 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-781 Supplier Abnova Corporation Supplier No. MAB9787
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against synthetic peptide of MT1A, MT1B, MT1E, MT1F, MT1G, MT1H, MT1IP, MT1L, MT1M, MT2A.

Specifications

Antigen MT1A, MT1B, MT1E, MT1F, MT1G, MT1H, MT1IP, MT1L, MT1M, MT2A
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone N11-G
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of MT1A, MT1B, MT1E, MT1F, MT1G, MT1H, MT1IP, MT1L, MT1M, MT2A.
Dilution Western Blot (1:1000) ELISA (1:5000-1:10000) The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris-HCl, pH 8.0 (10mg/mL BSA, 0.05% sodium azide)
Gene Accession No. P04731 (Gene ID : 4489);P07438 (Gene ID : 4490);P04732 (Gene ID : 4493);P04733 (Gene ID : 4494);P13640 (Gene ID : 4495);P80294 (Gene ID : 4496);P80295 (Gene ID : 4496);Q8N339 (Gene ID : 4499);Q86YX0 (Gene ID : 4490);Q86YX5 (Gene ID : 4489);Q8TDC4 (Gene ID
Gene Alias MGC32848/MT1/MT1S/MTC
Gene Symbols MT1A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human MT1A, MT1B, MT1E, MT1F, MT1G, MT1H, MT1IP, MT1L, MT1M, MT2A.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4489/4490/4493/4494/4495/4496/644314/4500/4499/450
Target Species Human, Mouse
Content And Storage Store at -20°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.