Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-MTHFR, Polyclonal,

Catalog No. ABS1013MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1013MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1013MI Supplier MilliporeSigma™ Supplier No. ABS1013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects MTHFR in Human, Mouse, Porcine samples, and it is validated for Western Blotting

Methylenetetrahydrofolate reductase or 5,10-methylenetetrahydrofolate reductase (EC 1.5.1.20; UniProt P42898) is encoded by the MTHFR gene (Gene ID 4524) in human. MTHFR catalyzes the reduction of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, the predominant circulatory form of folate and carbon donor for the re-methylation of homocysteine to methionine. Defects in MTHFR cause plasma homocysteine elevation or hperhomocysteinaemia, a risk factor for cerebrovascular, peripheral vascular, and coronary heart disease. Multiple transcription start sites and alternative splicing in the MTHFR gene result in two enzyme isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTHFR
Applications Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. P42898
Gene Symbols MTHFR
Host Species Rabbit
Immunogen Recombinant protein corresponding to human MTHFR.
Purification Method Unpurified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_005948
Target Species Human, Mouse, Pig
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.