Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTOR (phospho S2448), Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89114798 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-798 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-798 Supplier Abnova Corporation Supplier No. PAB10335
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of MTOR.

The protein encoded by this gene belongs to a family of phosphatidylinositol kinase-related kinases. These kinases mediate cellular responses to stresses such as DNA damage and nutrient deprivation. This protein acts as the target for the cell-cycle arrest and immunosuppressive effects of the FKBP12-rapamycin complex. The ANGPTL7 gene is located in an intron of this gene. [provided by RefSeq

Specifications

Antigen MTOR
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of MTOR.
Dilution ELISA (1:15000-1:65000) Western Blot (1:500-1:2000) Immunohistochemistry (5.0 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene MTOR
Gene Alias FRAP/FRAP1/FRAP2/RAFT1/RAPT1
Gene Symbols MTOR
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S2448 of human MTOR.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2475
Test Specificity Reactivity occurs with phosphorylated mTOR from human derived tissues and cells. Reactivity with mouse and rat is suggested based on protein sequence homologies.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.