Learn More
MUC2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MUC2.
Supplier: Abnova Corporation PAB14513
Description
This gene encodes a member of the mucin protein family. Mucins are high molecular weight glycoproteins produced by many epithelial tissues. The protein encoded by this gene is secreted and forms an insoluble mucous barrier that protects the gut lumen. The protein polymerizes into a gel of which 80% is composed of oligosaccharide side chains by weight. The protein features a central domain containing tandem repeats rich in threonine and proline that varies between 50 and 115 copies in different individuals. Alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known. [provided by RefSeq
Specifications
| MUC2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MUC2. | |
| In 10mM phosphate buffer, 150mM NaCl, pH 7.4 (1% BSA, 0.05% sodium azide) | |
| MLP/SMUC | |
| Rabbit | |
| 500 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) Immunohistochemistry (Frozen sections) (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| MUC2 | |
| MUC2 | |
| A synthetic peptide (conjugated with carrier protein) corresponding to human MUC2. | |
| RUO | |
| 4583 | |
| Store at 4°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only