Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mug1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116753 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-753 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-753 Supplier Abnova Corporation Supplier No. PAB13585
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against Mug1.

Specifications

Antigen Mug1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Mug1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.4
Gene Mug1
Gene Symbols Mug1
Host Species Rabbit
Immunogen Rat Mug1.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 497794
Target Species Rat
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 200μL deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.