Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MVK Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89131254 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-131-254 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-131-254 Supplier Abnova Corporation Supplier No. PAB4055
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of MVK.

This gene encodes the peroxisomal enzyme mevalonate kinase. Mevalonate is a key intermediate, and mevalonate kinase a key early enzyme, in isoprenoid and sterol synthesis. Mevalonate kinase deficiency caused by mutation of this gene results in mevalonic aciduria, a disease characterized psychomotor retardation, failure to thrive, hepatosplenomegaly, anemia and recurrent febrile crises. Defects in this gene also cause hyperimmunoglobulinaemia D and periodic fever syndrome, a disorder characterized by recurrent episodes of fever associated with lymphadenopathy, arthralgia, gastrointestinal dismay and skin rash. Two transcript variants that encode the same protein have been found for this gene. [provided by RefSeq

Specifications

Antigen MVK
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of MVK.
Dilution ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene MVK
Gene Alias FLJ96772/LRBP/MK/MVLK
Gene Symbols MVK
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human MVK.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4598
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.