Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYH9 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89117750 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-750 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-750 Supplier Abnova Corporation Supplier No. PAB15685
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Myh9.

Sequence: KKKQLVRQVREMEAELEDERKQRSMAMAARKKLEMDLKDLEAHIDTANKNREEAIKQLRKLQAQMKDCMRELDDTRASREEILAQAKENEKKLKSMEAEMIQLQEELAAAERAKRQAQQERDELADEIANSSGKGALALEEKRRLEARIAQLEEELEEEQGNTELINDRLKKANLQIDQINTDLNLERSHAQKNENARQQLERQNKELKAKLQEMESAVKSKYKASIAALEAKIAQLEEQLDNE

Specifications

Antigen MYH9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Myh9.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.02% sodium azide)
Gene Myh9
Gene Alias C80049/D0Jmb2/E030044M24Rik/Myhn-1/Myhn1/NMHCIIA/TU72.6
Gene Symbols Myh9
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to 244 mouse Myh9.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17886
Test Specificity Specific to recombinant protein GX0844. This antibody detects endogenous mMYH9 protein in several cell types. It also recognizes human MYH9 protein.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.