Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL6, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89134527 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-527 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-527 Supplier Abnova Corporation Supplier No. PAB9961
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of MYL6.

Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain that is expressed in smooth muscle and nonmuscle tissues. Genomic sequences representing several pseudogenes have been described and two transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq

Specifications

Antigen MYL6
Applications ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of MYL6.
Dilution ELISA (1:120000-1:135000) Western Blot (1:1000-1:5000) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene MYL6
Gene Alias ESMLC/LC17-GI/LC17-NM/LC17A/LC17B/MLC1SM/MLC3NM/MLC3SM
Gene Symbols MYL6
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S19/S20 of human MYL6.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4637
Test Specificity This antibody is phosphospecific and detect monophosphorylated and diphosphorylated forms of the protein. This phosphospecific polyclonal antibody is specific to the phosphorylated pS19/pS20 form of the protein, depending on the source origin of the protein.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.