Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYO5B Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89131870 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-131-870 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-131-870 Supplier Abnova Corporation Supplier No. PAB5468
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of MYO5B.

The protein encoded by this gene, together with other proteins, may be involved in plasma membrane recycling. Mutations in this gene are associated with microvillous inclusion disease. [provided by RefSeq

Specifications

Antigen MYO5B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of MYO5B.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene MYO5B
Gene Alias KIAA1119
Gene Symbols MYO5B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human MYO5B.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4645
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.