Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYOC, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89134059 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-059 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-059 Supplier Abnova Corporation Supplier No. PAB8971
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of MYOC.

MYOC encodes the protein myocilin, which is believed to have a role in cytoskeletal function. MYOC is expressed in many occular tissues, including the trabecular meshwork, and was revealed to be the trabecular meshwork glucocorticoid-inducible response protein (TIGR). The trabecular meshwork is a specialized eye tissue essential in regulating intraocular pressure, and mutations in MYOC have been identified as the cause of hereditary juvenile-onset open-angle glaucoma. [provided by RefSeq

Specifications

Antigen MYOC
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of MYOC.
Dilution Western Blot (1:500) Immunoprecipitation (1:200) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene MYOC
Gene Alias GLC1A/GPOA/JOAG/JOAG1/TIGR/myocilin
Gene Symbols MYOC
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human MYOC.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4653
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.