Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138108 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-108 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-108 Supplier Abnova Corporation Supplier No. PAB15563

Rabbit polyclonal antibody raised against full-length native Skeletal and Smooth Myosin.

Specifications

Antigen Myosin
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation In 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Host Species Rabbit
Immunogen Native purified bovine Skeletal and Smooth Myosin.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Immunofluorescent staining of cultured human and mouse fibroblasts shows a striated pattern of labeling along stress fibers. The specifically stains the A-band of skeletal muscle.
Target Species Bovine, Human, Mouse
Content And Storage Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.