Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-MYPT1, Polyclonal,

Catalog No. ABS1495MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1495MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1495MI Supplier MilliporeSigma™ Supplier No. ABS1495
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects MYPT1 in Human samples, and it is validated for Western Blotting

Chicken Protein phosphatase 1 regulatory subunit 12A (also known as PP1M subunit M110, Myosin phosphatase target subunit 1, Myosin phosphatase-targeting subunit 1, Protein phosphatase myosin-binding subunit, Smooth muscle myosin phosphatase myosin-binding subunit, Protein phosphatase 1, regulatory (inhibitor) subunit 12A, 130kDa myosin-binding subunit of smooth muscle myosin phophatase) is encoded by the Gallus gallus PP1R12A (also known as MBS, MBSP, MYPT1) gene (Entrez Gene ID 396020). MYTP1 protein is one of the subunits and an integral component of Myosin phosphatase. Myosin phosphatase regulates the interaction of Actin and Myosin downstream of the guanosine triphosphatase Rho, which inhibits Myosin phosphatase through the action of Rho-kinase. It is also involved in smooth muscle contraction. Inhibition of Myosin light chain phosphatase results in Ca2+ sensitization of smooth muscle contraction. This inhibition is modulated through phosphorylation of MYPT1 by a ZIP-like kinase, which associates with MYPTI and phosphorylates the inhibitory site in smooth muscle. The phosphorylation of MYPT1 by protein kinase C results in altered dephosphoryation of Myosin by attenuating the binding of protein phosphatase 1 catalytic subunit (PP1c) and the phosphorylated 20kDa Myosin light chain to MYPT1. PP1c interacts with at least four binding sites on the amino-terminus of MYPT1. MYPT1 is localized on stress fibers, and is distributed close to the cell membrane and at cell-cell contacts to regulate Myosin phosphatase activity. Anti-MYPT1 is raised against GST fustion protein containing recombinant chicken MYPT1 protein C-terminal tail fragment (UniProt Q90623) that is present in both isoform 1 (M133, 1004aa) and isoform 2 (M130, 963aa). Human reactivity of this product is confirmed by Western blotting analysis using HeLa cell lysate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYPT1
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q90623
Gene Symbols PPP1R12A; MBS; MYPT1
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to chicken MYPT1.
Purification Method Affinity Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_990454
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.