Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYST4, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116752 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-752 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-752 Supplier Abnova Corporation Supplier No. PAB13579
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of MYST4.

Specifications

Antigen MYST4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of MYST4.
Dilution Western Blot (1:7500) Immunohistochemistry (1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.4
Gene MYST4
Gene Symbols MYST4
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to bovine MYST4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 535403
Target Species Bovine
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 125μL deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.