Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myt1l, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Myt1l.
Supplier: Abnova Corporation PAB15836
Description
Sequence: RATSAMKKAKLSGEQMLTIKQRASNGIENDEEIKQLDEEIKELNESNSQMEADMIKLRTQITTMESNLKTIEEENKVIEQQNESLLHELANLSQSLIHSLANIQLPHMDPINEQNFDAYVTTLTEMYTNQDRYQSPENKALLENIKQAVRGIQVSpecifications
Myt1l | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant Myt1l. | |
In PBS (50% glycerol, 0.02% sodium azide) | |
2900046C06Rik/2900093J19Rik/C630034G21Rik/Nztf1/Pmng1/Png-1/mKIAA1106 | |
Rabbit | |
50 μg | |
Primary | |
Specific to recombinant protein GX0194. This antibody detects mMYT1L protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Myt1l | |
Myt1l | |
Recombinant GST fusion protein corresponding to 154 amino acids of mouse Myt1l. | |
RUO | |
17933 | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction