Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NANOG Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89116807 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-807 Supplier Abnova Corporation Supplier No. PAB13693
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against Nanog.

Specifications

Antigen NANOG
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Nanog.
Dilution Immunocytochemistry (1:150-1:700) Western Blot (1:300-1:2000) Immunoprecipitation (1:300) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Nanog
Gene Alias 2410002E02Rik/ENK/ecat4
Gene Symbols Nanog
Host Species Rabbit
Immunogen Mouse Nanog protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71950
Test Specificity Recognizes mouse Nanog. Cross reactivity with monkey and human observed in immunocytochemistry. Detects bands at 37-44 KDa.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.