Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAV3 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89124505 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-124-505 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-124-505 Supplier Abnova Corporation Supplier No. PAB22799
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant NAV3.

This gene belongs to the neuron navigator family and is expressed predominantly in the nervous system. The encoded protein contains coiled-coil domains and a conserved AAA domain characteristic for ATPases associated with a variety of cellular activities. This gene is similar to unc-53, a Caenorhabditis elegans gene involved in axon guidance. Multiple alternatively spliced transcript variants for this gene have been described but only one has had its full-length nature determined. [provided by RefSeq

Sequence: PSQSLSKPITMEKASASSCPAPLEGREAGQASPSGSCTMTVAQSSGQSTGNGAVQLPQQQQHSHPNTATVAPFIYRAHSENEGTALPSADS

Specifications

Antigen NAV3
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant NAV3.
Dilution Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene NAV3
Gene Alias KIAA0938/POMFIL1/STEERIN3/unc53H3
Gene Symbols NAV3
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human NAV3.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89795
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.