Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NBN Rabbit anti-Human, Mouse, Primates, Polyclonal Antibody, Abnova™

Catalog No. 89115752 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-752 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-752 Supplier Abnova Corporation Supplier No. PAB12377
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant NBN.

Mutations in this gene are associated with Nijmegen breakage syndrome, an autosomal recessive chromosomal instability syndrome characterized by microcephaly, growth retardation, immunodeficiency, and cancer predisposition. The encoded protein is a member of the MRE11/RAD50 double-strand break repair complex which consists of 5 proteins. This gene product is thought to be involved in DNA double-strand break repair and DNA damage-induced checkpoint activation. [provided by RefSeq

Specifications

Antigen NBN
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant NBN.
Dilution Immunocytochemistry (1:50-1:200) Immunofluorescence (1:200) Immunoprecipitation (3 ug/mL) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.09% sodium azide)
Gene NBN
Gene Alias AT-V1/AT-V2/ATV/FLJ10155/MGC87362/NBS/NBS1/P95
Gene Symbols NBN
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1-705 of human NBN.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4683
Test Specificity This antibody is specific to human NBS1.
Target Species Human, Mouse, Primate
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.