Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAM2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89113972 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-972 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-972 Supplier Abnova Corporation Supplier No. PAB0134
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against mouse NCAM-2 (442 a.a.-685 a.a) partial recombinant protein.

Specifications

Antigen NCAM2
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against mouse NCAM-2 (442 a.a. ∼ 685 a.a) partial recombinant protein.
Dilution Western Blot (1:1000) Immunohistochemistry (1:500) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.4
Gene Ncam2
Gene Alias Ncam-2/Ocam/RNCAM
Gene Symbols Ncam2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 442-685 of Mouse NCAM-2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17968
Target Species Mouse
Content And Storage Store at -20°C on dry atmosphere.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.