Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCL Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89115751 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-751 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-751 Supplier Abnova Corporation Supplier No. PAB12376
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant NCL.

Nucleolin (NCL), a eukaryotic nucleolar phosphoprotein, is involved in the synthesis and maturation of ribosomes. It is located mainly in dense fibrillar regions of the nucleolus. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. The intron 11 of the NCL gene encodes a small nucleolar RNA, termed U20. [provided by RefSeq

Specifications

Antigen NCL
Applications Immunofluorescence, Immunohistochemistry, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant NCL.
Dilution Immunofluorescence (1:50-1:100) Western Blot (1:4000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.09% sodium azide)
Gene NCL
Gene Alias C23/FLJ45706
Gene Symbols NCL
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 284-709 of human NCL.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4691
Test Specificity This antibody is specific to nucleolin protein.
Target Species Human, Mouse
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.