Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCOA3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114753 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-753 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-753 Supplier Abnova Corporation Supplier No. PAB10282
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of NCOA3.

The protein encoded by this gene is a nuclear receptor coactivator that interacts with nuclear hormone receptors to enhance their transcriptional activator functions. The encoded protein has histone acetyltransferase activity and recruits p300/CBP-associated factor and CREB binding protein as part of a multisubunit coactivation complex. This protein is initially found in the cytoplasm but is translocated into the nucleus upon phosphorylation. Two transcript variants encoding different isoforms have been found for this gene. In addition, a polymorphic repeat region is found in the C-terminus of the encoded protein. [provided by RefSeq

Specifications

Antigen NCOA3
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NCOA3.
Dilution ELISA (1:175000) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene NCOA3
Gene Alias ACTR/AIB-1/AIB1/CAGH16/CTG26/KAT13B/MGC141848/RAC3/SRC3/TNRC14/TNRC16/TRAM-1/bHLHe42/pCIP
Gene Symbols NCOA3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region of human NCOA3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8202
Test Specificity This antibody reacts with several isoforms of human NCOA3 protein (a, b, d, e and f).
Target Species Bovine, Canine, Human, Macaque, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.