Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCR3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89119459 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-459 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-459 Supplier Abnova Corporation Supplier No. PAB17794
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of NCR3.

Natural cytotoxicity receptors (NCRs), such as NCR3, are activating natural killer (NK) cell receptors that belong to the immunoglobulin (Ig) superfamily. NCR3 is expressed in all resting and activated NK cells and forms a complex with CD3-zeta (CD3Z, or CD247; MIM 186780) (Sato et al., 2001 [PubMed 11782277]).[supplied by OMIM

Specifications

Antigen NCR3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NCR3.
Dilution Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Gene NCR3
Gene Alias 1C7/CD337/LY117/MALS/NKp30
Gene Symbols NCR3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal of human NCR3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259197
Test Specificity This antibody detects endogenous levels of total NCR3 protein
Target Species Human, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.