Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFA13 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114837 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-837 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-837 Supplier Abnova Corporation Supplier No. PAB1041

Rabbit polyclonal antibody raised against synthetic peptide of NDUFA13.

Sequence: QAETDRRTLQMLRE

Specifications

Antigen NDUFA13
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13
Formulation In PBS (0.02% sodium azide)
Gene NDUFA13
Gene Alias B16.6, CDA016, CGI-39, GRIM-19, GRIM19
Gene Symbols NDUFA13
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 159-172 of human NDUFA13.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51079
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.