Learn More
NDUFB7, beta subcomplex 1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NDUFB7, beta subcomplex 1.
Supplier: Abnova Corporation PAB28689
Description
The protein encoded by this gene is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It is located at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. [provided by RefSeq
Sequence: FPERKEREMVATQQEMMDAQLRLQLRDYCAHHLIRLLKCKRDSFPNFLACKQERHDWDYCEHRDYVMRMKEFERERRLLQRKKRREKKAAELAKGQGPGEVDPKVSpecifications
NDUFB7, beta subcomplex 1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant NDUFB7, beta subcomplex 1. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
B18/CI-B18/MGC2480 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
4713 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Immunohistochemistry (1:20-1:50) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. | |
NDUFB7 | |
NDUFB7 | |
Recombinant protein corresponding to amino acids of human NDUFB7. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only