Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEFH Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114512 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-512 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-512 Supplier Abnova Corporation Supplier No. PAB0953

Rabbit polyclonal antibody raised against synthetic peptide of NEFH.

Neurofilaments are type IV intermediate filament heteropolymers composed of light, medium, and heavy chains. Neurofilaments comprise the axoskeleton and functionally maintain neuronal caliber. They may also play a role in intracellular transport to axons and dendrites. This gene encodes the heavy neurofilament protein. This protein is commonly used as a biomarker of neuronal damage and susceptibility to amyotrophic lateral sclerosis (ALS) has been associated with mutations in this gene. [provided by RefSeq

Sequence: SPEKAKSPEKAKSC

Specifications

Antigen NEFH
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description neurofilament, heavy polypeptide
Formulation In PBS (0.02% sodium azide)
Gene NEFH
Gene Alias NFH
Gene Symbols NEFH
Host Species Rabbit
Immunogen A synthetic peptide corresponding to repeated motif, XKSPYK domain of human NEFH.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4744
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.