Learn More
NEIL3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NEIL3.
Supplier: Abnova Corporation PAB1054
Description
NEIL3 belongs to a class of DNA glycosylases homologous to the bacterial Fpg/Nei family. These glycosylases initiate the first step in base excision repair by cleaving bases damaged by reactive oxygen species and introducing a DNA strand break via the associated lyase reaction (Bandaru et al., 2002 [PubMed 12509226]).[supplied by OMIM
Sequence: LRAESEVKKQKGRMLGSpecifications
NEIL3 | |
Polyclonal | |
nei endonuclease VIII-like 3 (E. coli) | |
NEIL3 | |
NEIL3 | |
A synthetic peptide corresponding to amino acids 164-177 of human NEI3. | |
RUO | |
55247 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
In PBS (0.02% sodium azide) | |
FGP2, FLJ10858, FPG2, NEI3, hFPG2, hNEI3 | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only