Learn More
NEK3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NEK3.
Supplier: Abnova Corporation PAB3285
Description
This gene encodes a member of the NimA (never in mitosis A) family of serine/threonine protein kinases. The encoded protein differs from other NimA family members in that it is not cell cycle regulated and is found primarily in the cytoplasm. The kinase is activated by prolactin stimulation, leading to phosphorylation of VAV2 guanine nucleotide exchange factor, paxillin, and activation of the RAC1 GTPase. Multiple transcript variants encoding different isoforms have been found for this gene
Specifications
| NEK3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NEK3. | |
| In PBS (0.09% sodium azide) | |
| HSPK36/MGC29949 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 4752 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| NEK3 | |
| NEK3 | |
| A synthetic peptide (conjugated with KLH) corresponding to internal region of human NEK3. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only