Learn More
Specifications
Specifications
| Antigen | Nephrophthisis (pic) |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against partial recombinant Nephrophthisis (pic). |
| Dilution | Western blot (2 to 10 ug/mL) Immunoprecipitation (10 to 20 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS (0.08% sodium azide) |
| Host Species | Rabbit |
| Immunogen | Recombinant GST fusion protein corresponding to amino acid 612-691 of mouse pic. |
| Quantity | 250 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.