Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NES, Rabbit, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89117397 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-397 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-397 Supplier Abnova Corporation Supplier No. PAB14991
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant NES.

Nestin is an intermediate filament protein that was first identified with a monoclonal antibody by Hockfield and McKay (1985) [PubMed 4078630]. It is expressed predominantly in stem cells of the central nervous system in the neural tube. Upon terminal neural differentiation, nestin is downregulated and replaced by neurofilaments.[supplied by OMIM

Specifications

Antigen NES
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Description Rabbit polyclonal antibody raised against recombinant NES.
Dilution Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene NES
Gene Alias FLJ21841/Nbla00170
Gene Symbols NES
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1464-1614 of human NES.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10763
Test Specificity It does not cross-react with rodent protein. This is specific to human nestin. This antibody may be used in Western blot where a band is seen at ∼220-240 KDa. Another band may be seen ∼110-115 KDa.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.