Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NES, Rabbit, Clone: SP103, Abnova™

Catalog No. 89108366 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-366 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-366 Supplier Abnova Corporation Supplier No. MAB10050

Rabbit monoclonal antibody raised against synthetic peptide of NES.

Nestin is an intermediate filament protein that was first identified with a monoclonal antibody by Hockfield and McKay (1985) [PubMed 4078630]. It is expressed predominantly in stem cells of the central nervous system in the neural tube. Upon terminal neural differentiation, nestin is downregulated and replaced by neurofilaments.[supplied by OMIM

Specifications

Antigen NES
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SP103
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of NES.
Dilution Western Blot (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene NES
Gene Alias FLJ21841/Nbla00170
Gene Symbols NES
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human NES.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10763
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.