Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEU2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121446 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-446 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-446 Supplier Abnova Corporation Supplier No. PAB19939
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of NEU2.

This gene belongs to a family of glycohydrolytic enzymes which remove sialic acid residues from glycoproteins and glycolipids. Expression studies in COS7 cells confirmed that this gene encodes a functional sialidase. Its cytosolic localization was demonstrated by cell fractionation experiments. [provided by RefSeq

Specifications

Antigen NEU2
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NEU2.
Dilution ELISA (1:10000-1:50000) Western Blot (1:500- 1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene NEU2
Gene Alias MGC129579/SIAL2
Gene Symbols NEU2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 110-124 of human NEU2.
Purification Method Immunoaffinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4759
Test Specificity This antibody reacts with human Neu2. Based on sequence we expect this antibody to react with neuraminidase from other sources, although specific reactivity has not been confirmed. Cross-reactivity against Neu1 has not yet been established. Neuraminidases are highly conserved in mammals and therefore cross reactivity is expected with mouse and rat Neu2.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.