Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEURL Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NEURL.
Supplier: Abnova Corporation PAB24309
Description
Sequence: PDLVSQSGFWAKALPEEFANEGNIIAFWVDKKGRVFHRINDSAVMLFFSGVRTADPLWALVDSpecifications
NEURL | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant NEURL. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
NEURL1/RNF67/h-neu/neu-1 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
9148 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
NEURL | |
NEURL | |
Recombinant protein corresponding to amino acids of human NEURL. | |
100 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction