Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofibrillary Tangle, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137801 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-801 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-801 Supplier Abnova Corporation Supplier No. PAB14018

Rabbit polyclonal antibody raised against Neurofibrillary Tangle.

Specifications

Antigen Neurofibrillary Tangle
Applications ELISA, Immunodiffusion, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Neurofibrillary Tangle.
Dilution ELISA (>1:4000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (>1:200) Immunohistochemistry (Frozen sections) (>1:2000) Western Blot (>1:500) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Host Species Rabbit
Immunogen Disaggregated paired helical filaments.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity human paired helical filaments from neurofibrillary tangles. Reacts with both intracellular and extracellular neurofibrillary tangles.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.