Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGFR Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89117001 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-001 Supplier Abnova Corporation Supplier No. PAB14320
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial NGFR.

Nerve growth factor receptor contains an extracellular domain containing four 40-amino acid repeats with 6 cysteine residues at conserved positions followed by a serine/threonine-rich region, a single transmembrane domain, and a 155-amino acid cytoplasmic domain. The cysteine-rich region contains the nerve growth factor binding domain. [provided by RefSeq

Specifications

Antigen NGFR
Applications Immunofluorescence, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial NGFR.
Dilution Immunohistochemistry (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene NGFR
Gene Alias CD271/Gp80-LNGFR/TNFRSF16/p75(NTR)/p75NTR
Gene Symbols NGFR
Host Species Rabbit
Immunogen Human extracellular domain NGFR.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4804
Test Specificity IHC shows specific staining for p75NTR.
Target Species Human, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.