Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIPSNAP3B, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89118512 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-512 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-512 Supplier Abnova Corporation Supplier No. PAB16677
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of NIPSNAP3B.

NIPSNAP3B belongs to a family of proteins with putative roles in vesicular trafficking (Buechler et al., 2004 [PubMed 15177564]).[supplied by OMIM

Specifications

Antigen NIPSNAP3B
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NIPSNAP3B.
Dilution Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene NIPSNAP3B
Gene Alias FLJ11275/NIPSNAP3
Gene Symbols NIPSNAP3B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region 16 amino acids of human NIPSNAP3B.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55335
Target Species Human, Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.