Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKIRAS1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NKIRAS1.
Supplier: Abnova Corporation PAB12869
Specifications
NKIRAS1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of NKIRAS1. | |
In PBS (0.02% sodium azide) | |
KBRAS1/kappaB-Ras1 | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
NKIRAS1 | |
NKIRAS1 | |
A synthetic peptide corresponding to internal region 18 amino acids of human NKIRAS1. | |
RUO | |
28512 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction