Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKIRAS1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89114728 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-728 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-728 Supplier Abnova Corporation Supplier No. PAB10253
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of NKIRAS1.

Sequence: LPGRKNKGNSNSEN

Specifications

Antigen NKIRAS1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NKIRAS1.
Dilution ELISA (1:10000-1:50000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene NKIRAS1
Gene Alias KBRAS1/kappaB-Ras1
Gene Symbols NKIRAS1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 179-192 of human NKIRAS1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28512
Test Specificity A single amino acid changes of S190P exists in mouse.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.