Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX2-1, Rabbit, Clone: G21-G, Abnova™

Catalog No. 89113777 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-777 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-777 Supplier Abnova Corporation Supplier No. MAB9783
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against synthetic peptide of NKX2-1.

Specifications

Antigen NKX2-1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone G21-G
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of NKX2-1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris-HCl, pH 8.0 (20mg/mL BSA, 0.05% sodium azide)
Gene NKX2-1
Gene Accession No. P43699
Gene Alias BCH/BHC/NK-2/NKX2.1/NKX2A/TEBP/TITF1/TTF-1/TTF1
Gene Symbols NKX2-1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human NKX2-1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7080
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.